missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-2-phenyl-1H-inden-1-one, tech., Thermo Scientific™

Product Code. 10190732
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10190732 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10190732 Supplier Thermo Scientific Maybridge Supplier No. CD09182DA

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 1947-47-3
CAS Min % 80.0
CAS Max % 100.0
Assay Percent Range 90%
Packaging Amber Glass Bottle
Molecular Formula C15H11NO
MDL Number MFCD00051614
Quantity 1 g
Synonym 3-amino-2-phenyl-1h-inden-1-one, 3-amino-2-phenylindenone, maybridge1_003093, acmc-1c032, 3-amino-2-phenyl-inden-1-one, 2-phenyl-3-amino-1h-indene-1-one, 1h-inden-1-one,3-amino-2-phenyl, 3-amino-2-phenyl-1h-inden-1-one, tech.
InChI Key HLEKBTIHDXNOQP-UHFFFAOYSA-N
SMILES C1=CC=C(C=C1)C2=C(C3=CC=CC=C3C2=O)N
IUPAC Name 3-amino-2-phenylinden-1-one
Molecular Weight (g/mol) 221.259
PubChem CID 291219
Formula Weight 221.26
Percent Purity Tech.
Grade Technical
Chemical Name or Material 3-Amino-2-phenyl-1H-inden-1-one
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.