missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Amino-2-naphthoic acid, 97%
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
Beschreibung
3-Amino-2-naphthoic acid is a dyes analytical reagent which is used as an intermediate for organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-Amino-2-naphthoic acid is a dyes analytical reagent which is used as an intermediate for organic synthesis.
Solubility
Soluble in alcohol and ether. Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
3-Amino-2-naphthoic acid is a dyes analytical reagent which is used as an intermediate for organic synthesis.
Solubility
Soluble in alcohol and ether. Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
Spezifikation
Spezifikation
| Melting Point | ∽210°C (decomposition) |
| Quantity | 1 g |
| Beilstein | 744099 |
| Merck Index | 14,452 |
| Solubility Information | Soluble in alcohol and ether. Insoluble in water. |
| SMILES | C1=CC=C2C=C(C(=CC2=C1)C(=O)O)N |
| Molecular Weight (g/mol) | 187.198 |
| Formula Weight | 187.2 |
| Percent Purity | 97% |
| Chemical Name or Material | 3-Amino-2-naphthoic acid |
RUO – Research Use Only
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur