Learn More
3-Amino-2-naphthoic acid, 85%, tech., Thermo Scientific Chemicals
£74.30 - £279.00
Chemical Identifiers
CAS | 5959-52-4 |
---|---|
Molecular Formula | C11H9NO2 |
Molecular Weight (g/mol) | 187.2 |
MDL Number | MFCD00004115 |
InChI Key | XFXOLBNQYFRSLQ-UHFFFAOYSA-N |
Synonym | 3-amino-2-naphthoic acid, 2-amino-3-naphthoic acid, 3-aminoisonaphthoic acid, 2-naphthalenecarboxylic acid, 3-amino, 2-amino-3-carboxynaphthalene, 2-naphthoic acid, 3-amino, 3-amino-2-naphthalenecarboxylic acid, unii-iv5z0phl5d, iv5z0phl5d, 3-amino-naphthalene-2-carboxylic acid |
PubChem CID | 22244 |
IUPAC Name | 3-aminonaphthalene-2-carboxylic acid |
SMILES | C1=CC=C2C=C(C(=CC2=C1)C(=O)O)N |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10071841
|
Thermo Scientific Acros
104140050 |
5 g | Glass bottle |
£74.30
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10001871
|
Thermo Scientific Acros
104140250 |
25 g | Glass bottle |
£279.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
5959-52-4 | |
187.2 | |
XFXOLBNQYFRSLQ-UHFFFAOYSA-N | |
22244 | |
C1=CC=C2C=C(C(=CC2=C1)C(=O)O)N |
C11H9NO2 | |
MFCD00004115 | |
3-amino-2-naphthoic acid, 2-amino-3-naphthoic acid, 3-aminoisonaphthoic acid, 2-naphthalenecarboxylic acid, 3-amino, 2-amino-3-carboxynaphthalene, 2-naphthoic acid, 3-amino, 3-amino-2-naphthalenecarboxylic acid, unii-iv5z0phl5d, iv5z0phl5d, 3-amino-naphthalene-2-carboxylic acid | |
3-aminonaphthalene-2-carboxylic acid |
Specifications
5959-52-4 | |
100.0 | |
Green to Green-Yellow | |
85% | |
H2NC10H6CO2H | |
14, 535 | |
3-amino-2-naphthoic acid, 2-amino-3-naphthoic acid, 3-aminoisonaphthoic acid, 2-naphthalenecarboxylic acid, 3-amino, 2-amino-3-carboxynaphthalene, 2-naphthoic acid, 3-amino, 3-amino-2-naphthalenecarboxylic acid, unii-iv5z0phl5d, iv5z0phl5d, 3-amino-naphthalene-2-carboxylic acid | |
C1=CC=C2C=C(C(=CC2=C1)C(=O)O)N | |
187.2 | |
187.2 | |
Technical | |
3-Amino-2-naphthoic acid, 85% |
80.0 | |
210.0°C | |
Authentic | |
C11H9NO2 | |
MFCD00004115 | |
15, 448 | |
XFXOLBNQYFRSLQ-UHFFFAOYSA-N | |
3-aminonaphthalene-2-carboxylic acid | |
22244 | |
85% | |
Powder |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 227-726-2
missing translation for 'rtecsNumber' : QL1400000
missing translation for 'tsca' : TSCA
- 3-Amino-2-naphthoic acid, tech.
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H302-Harmful if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only