missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-2-methylbenzoic acid, 98%

Product Code. 11481477
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11481477 5 g 5g
11491477 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11481477 Supplier Thermo Scientific Alfa Aesar Supplier No. B24947.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 52130-17-3 | C8H9NO2 | 151.165 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52130-17-3
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.165
MDL Number MFCD00075026
InChI Key BYHMLZGICSEKIY-UHFFFAOYSA-N
Synonym 2-methyl-3-aminobenzoic acid, 3-amino-o-toluic acid, 3-amino-2-methylbenzoicacid, 3-amino-2-methyl-benzoic acid, benzoic acid, 3-amino-2-methyl, 2-methyl-3-amino benzoic acid, pubchem10921, acmc-209ky1, ksc489o4b, 3-amino-2-methyl benzoic acid
PubChem CID 2733699
IUPAC Name 3-amino-2-methylbenzoic acid
SMILES CC1=C(C=CC=C1N)C(=O)O

Specifications

Melting Point 185°C to 188°C
Quantity 5 g
Beilstein 2803090
Formula Weight 151.16
Percent Purity 98%
Chemical Name or Material 3-Amino-2-methylbenzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.