missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-2-methoxypyridine-5-boronic acid pinacol ester, 96%, Thermo Scientific Chemicals

Product Code. 15401356
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15401356 1 g 1g
15411356 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15401356 Supplier Thermo Scientific Alfa Aesar Supplier No. H54570.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

It is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Incompatible with oxidizing agents, air, heat. Refrigerated (2-4°C).
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 893440-50-1
Molecular Formula C12H19BN2O3
Molecular Weight (g/mol) 250.11
MDL Number MFCD12923427
InChI Key KYYKGOURQXPERA-UHFFFAOYSA-N
Synonym 2-methoxy-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-3-amine, 3-amino-2-methoxypyridine-5-boronic acid pinacol ester, 5-amino-6-methoxypyridine-3-boronic acid pinacol ester, 2-methoxy-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-pyridin-3-ylamine, 2-methoxy-3-aminopyridine-5-boronic acid pinacol ester, 5-amino-6-methoxypyridin-3-yl boronic acid pinacol ester, 2-methoxy-5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-3-amine, 5-amino-6-methoxypyridine-3-boronic acid pinacol ester?, 3-amino-2-methoxypyridin-5-ylboronicacidpinacolester, 5-amino-6-methoxypyridine-3-boronicacidpinacolester
PubChem CID 59557920
IUPAC Name 2-methoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-3-amine
SMILES COC1=C(N)C=C(C=N1)B1OC(C)(C)C(C)(C)O1

Specifications

Melting Point 131°C to 133°C
Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 250.1
Percent Purity 96%
Chemical Name or Material 3-Amino-2-methoxypyridine-5-boronic acid pinacol ester

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.