missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-2-fluoro-propanoic Acid Hydrochloride, TRC

Product Code. 30396897
Change view
Click to view available options
Quantity:
25 mg
100 mg
250 mg
Unit Size:
100mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30396897 100 mg 100mg
30401392 25 mg -
30394864 250 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30396897 Supplier Toronto Research Chemicals Supplier No. TRCF587355100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 867-84-5
Molecular Formula C3H7ClFNO2
Molecular Weight (g/mol) 143.54
Synonym 2-Fluoro-β-alanine Hydrochloride
SMILES Cl.NCC(F)C(=O)O

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C3H6FNO2.ClH/c4-2(1-5)3(6)7;/h2H,1,5H2,(H,6,7);1H
Chemical Name or Material 3-Amino-2-fluoro-propanoic Acid Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.