missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-2-cyclohexen-1-one, 98% (dry wt.), may cont. up to 5% water

Product Code. 11440363
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11440363 1 g 1g
11450363 5 g 5g
11460363 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11440363 Supplier Thermo Scientific Alfa Aesar Supplier No. A11474.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 5220-49-5 | C6H9NO | 111.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5220-49-5
Molecular Formula C6H9NO
Molecular Weight (g/mol) 111.14
MDL Number MFCD00013783
InChI Key ZZMRPOAHZITKBV-UHFFFAOYSA-N
Synonym 3-amino-2-cyclohexen-1-one, 3-aminocyclohex-2-enone, 2-cyclohexen-1-one, 3-amino, unii-v596547zke, 3-amino-2-cyclohexen-1-ketone, 3-amino-2-cyclohexenone, 3-aminocyclohexenone, 3-aminocyclohex-enone, pubchem14449, acmc-209kyd
PubChem CID 78892
IUPAC Name 3-aminocyclohex-2-en-1-one
SMILES NC1=CC(=O)CCC1

Specifications

Melting Point 128°C to 133°C (dried)
Quantity 1 g
Beilstein 471354
Formula Weight 111.15
Percent Purity 98%
Concentration or Composition (by Analyte or Components) May contain up to 5% water
Assay (dry wt.)
Chemical Name or Material 3-Amino-2-cyclohexen-1-one

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.