missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-2-cyclohexen-1-one, 95%, Thermo Scientific™

Product Code. 15284684 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15284684 5 g Glass bottle 5g
15294684 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15284684 Supplier Thermo Scientific Acros Supplier No. 219310050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 5220-49-5
Molecular Formula C6H9NO
Molecular Weight (g/mol) 111.14
MDL Number MFCD00013783
InChI Key ZZMRPOAHZITKBV-UHFFFAOYSA-N
Synonym 3-amino-2-cyclohexen-1-one, 3-aminocyclohex-2-enone, 2-cyclohexen-1-one, 3-amino, unii-v596547zke, 3-amino-2-cyclohexen-1-ketone, 3-amino-2-cyclohexenone, 3-aminocyclohexenone, 3-aminocyclohex-enone, pubchem14449, acmc-209kyd
PubChem CID 78892
IUPAC Name 3-aminocyclohex-2-en-1-one
SMILES NC1=CC(=O)CCC1

Specifications

Melting Point 133.0°C to 135.0°C
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 111.14
Percent Purity 95%
Chemical Name or Material 3-Amino-2-cyclohexen-1-one
compliance-icons
Product Identifier
  • 3-Amino-2-cyclohexen-1-one
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.