missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-2-chloropyridine, 98+%

Product Code. 11472508
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11472508 10 g 10g
11482508 50 g 50g
11492508 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11472508 Supplier Thermo Scientific Alfa Aesar Supplier No. A15830.09
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 6298-19-7 | C5H5ClN2 | 128.56 g/mol

It is an intermediate of medicine and agricultural chemicals. It is involved in the Suzuki-Miyaura cross-coupling reactions of heteroaryl compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an intermediate of medicine and agricultural chemicals. It is involved in the Suzuki-Miyaura cross-coupling reactions of heteroaryl compounds.

Solubility
Soluble in methanol and water. (30 g/L).

Notes
Store at room temperature. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6298-19-7
Molecular Formula C5H5ClN2
Molecular Weight (g/mol) 128.56
MDL Number MFCD00006238
InChI Key MEQBJJUWDCYIAB-UHFFFAOYSA-N
Synonym 3-amino-2-chloropyridine, 2-chloro-3-aminopyridine, 2-chloro-3-pyridinamine, 3-pyridinamine, 2-chloro, 2-chloro-3-amino pyridine, 2-chloro-3-pyridylamine, 2-chloro-3-amino-pyridine, 2-chloro-pyridin-3-ylamine, 2-chloro-3-pyridinylamine, 3-amino-2-chloro-pyridine
PubChem CID 80528
IUPAC Name 2-chloropyridin-3-amine
SMILES NC1=CC=CN=C1Cl

Specifications

Melting Point 76°C to 80°C
Boiling Point 130°C to 134°C (22 mmHg)
Flash Point 185°C (365°F)
Odor Unpleasant
Quantity 10 g
Beilstein 109814
Solubility Information Soluble in methanol and water. (30g/L).
Formula Weight 128.56
Percent Purity ≥98%
Chemical Name or Material 3-Amino-2-chloropyridine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.