missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-2-chloropyridine, 96%

Product Code. 10616862 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10616862 10 g Glass bottle 10g
10626862 50 g Glass bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10616862 Supplier Thermo Scientific Acros Supplier No. 103420100
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 6298-19-7 | C5H5ClN2 | 128.56 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6298-19-7
Molecular Formula C5H5ClN2
Molecular Weight (g/mol) 128.56
MDL Number MFCD00006238
InChI Key MEQBJJUWDCYIAB-UHFFFAOYSA-N
Synonym 3-amino-2-chloropyridine, 2-chloro-3-aminopyridine, 2-chloro-3-pyridinamine, 3-pyridinamine, 2-chloro, 2-chloro-3-amino pyridine, 2-chloro-3-pyridylamine, 2-chloro-3-amino-pyridine, 2-chloro-pyridin-3-ylamine, 2-chloro-3-pyridinylamine, 3-amino-2-chloro-pyridine
PubChem CID 80528
IUPAC Name 2-chloropyridin-3-amine
SMILES NC1=CC=CN=C1Cl

Specifications

CAS Min % 95.0
CAS Max % 100.0
Melting Point 77.0°C to 82.0°C
Color Pink to White
Infrared Spectrum Authentic
Assay Percent Range 95% min. (HPLC)
Packaging Glass bottle
Linear Formula Cl(C5H3N)NH2
Quantity 10 g
Formula Weight 128.56
Percent Purity 96%
Physical Form Crystalline Powder, Crystals or Chunks
Chemical Name or Material 3-Amino-2-chloropyridine, 96%
Show More Show Less
compliance-icons
Product Identifier
  • 3-Amino-2-chloropyridine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.