missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-Amino-2-chloro-6-methylpyridine, 98%

Product Code. 15401189
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15401189 1 g 1g
15411189 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15401189 Supplier Alfa Aesar™ Supplier No. H64125.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 39745-40-9
Molecular Formula C6H7ClN2
Molecular Weight (g/mol) 142.59
MDL Number MFCD03095213
InChI Key SUMSBUDMYMYTLL-UHFFFAOYSA-N
Synonym 3-amino-2-chloro-6-picoline, 3-amino-2-chloro-6-methylpyridine, 5-amino-6-chloro-2-picoline, 2-chloro-6-methyl-pyridin-3-ylamine, 2-chloro-3-amino-6-methylpyridine, 3-pyridinamine, 2-chloro-6-methyl, 2-chloro-6-methyl-3-pyridylamine, pubchem1245, acmc-20dld6, 2-chloro-3-amino-6-picoline
PubChem CID 2734415
IUPAC Name 2-chloro-6-methylpyridin-3-amine
SMILES CC1=CC=C(N)C(Cl)=N1

Specifications

Melting Point 64°C to 72°C
Color White to Yellow
Quantity 1 g
UN Number UN2811
Beilstein 114030
Formula Weight 142.59
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 3-Amino-2-chloro-6-methylpyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.