missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-2-bromopyridine, 97%

Product Code. 11359825
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11359825 25 g 25g
11339825 1 g 1g
11349825 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11359825 Supplier Thermo Scientific Alfa Aesar Supplier No. L19839.14
 Restricted Item
Estimated Shipment: 16-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 39856-58-1 | C5H5BrN2 | 173.013 g/mol

Used in a palladium-catalyzed synthesis of indole systems containing fused medium- and large-ring heterocycles. pyrrole derivatives ahs been prepared using 3-Amino-2-bromopyridine. Used in P-C coupling reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used in a palladium-catalyzed synthesis of indole systems containing fused medium- and large-ring heterocycles. pyrrole derivatives ahs been prepared using 3-Amino-2-bromopyridine. Used in P-C coupling reaction.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39856-58-1
Molecular Formula C5H5BrN2
Molecular Weight (g/mol) 173.013
MDL Number MFCD00234064
InChI Key HKDVVTLISGIPFE-UHFFFAOYSA-N
Synonym 3-amino-2-bromopyridine, 2-bromo-3-aminopyridine, 2-bromo-pyridineamine, 3-pyridinamine, 2-bromo, 2-bromo-3-pyridinamine, 2-bromo-3-pyridylamine, 2-bromopyridine-3-amine, 3-amino-2-bromo-pyridine, 2-bromo-3-pyridineamine, 2-bromo-pyridin-3-ylamine
PubChem CID 642816
IUPAC Name 2-bromopyridin-3-amine
SMILES C1=CC(=C(N=C1)Br)N

Specifications

Melting Point 75°C to 79°C
Quantity 25 g
Beilstein 109830
Solubility Information Slightly soluble in water.
Formula Weight 173.01
Percent Purity 98%
Chemical Name or Material 3-Amino-2-bromopyridine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.