missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-2-bromo-6-methylpyridine, 98%

Product Code. 15471439
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15471439 1 g 1g
15491439 250 mg 250mg
15481439 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15471439 Supplier Thermo Scientific Alfa Aesar Supplier No. H64301.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 126325-53-9 | C6H7BrN2 | 187.04 g/mol

It is an important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.

Solubility
Slightly soluble in water.

Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 126325-53-9
Molecular Formula C6H7BrN2
Molecular Weight (g/mol) 187.04
MDL Number MFCD16606103
InChI Key ORELOTUWTIKRIW-UHFFFAOYSA-N
Synonym 3-amino-4-bromo-2-methylpyridine, ksc915a1f, 3-pyridinamine, 4-bromo-2-methyl, 4-bromo-2-methyl-3-pyridinamine
PubChem CID 14418044
IUPAC Name 4-bromo-2-methylpyridin-3-amine
SMILES CC1=NC=CC(Br)=C1N

Specifications

Color White
Quantity 1 g
UN Number UN2811
Beilstein 4243986
Solubility Information Slightly soluble in water.
Formula Weight 187.04
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 3-Amino-2-bromo-6-methylpyridine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.