missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-2,2-dimethyl-1-propanol, 95%

Product Code. 11398425
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11398425 1 g 1g
11308435 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11398425 Supplier Thermo Scientific Alfa Aesar Supplier No. L10429.03
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 26734-09-8 | C5H13NO | 103.165 g/mol

Reatant in the synthesis of cyclic carbamates, cryptophycin analogues. Neopentanolamine with ammonia and hydrogen yeilds neopentyldiamine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reatant in the synthesis of cyclic carbamates, cryptophycin analogues. Neopentanolamine with ammonia and hydrogen yeilds neopentyldiamine.

Solubility
Soluble in Dichloromethane, Chloroforn, Ether and Methanol

Notes
Hygroscopic,Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26734-09-8
Molecular Formula C5H13NO
Molecular Weight (g/mol) 103.165
MDL Number MFCD00059847
InChI Key FNVOFDGAASRDQY-UHFFFAOYSA-N
Synonym 3-amino-2,2-dimethyl-1-propanol, neopentanolamine, 1-propanol, 3-amino-2,2-dimethyl, 3-amino-2,2-dimethylpropanol, 2,2-dimethyl-3-aminopropanol, 1-amino-2,2-dimethyl-3-propanol, 3-amino-2,2-dimethyl-propan-1-ol, neo-pentanolamine, acmc-1cdms, ksc496k6d
PubChem CID 117326
IUPAC Name 3-amino-2,2-dimethylpropan-1-ol
SMILES CC(C)(CN)CO

Specifications

Melting Point ∼70°C
Quantity 1 g
Beilstein 1731900
Sensitivity Hygroscopic
Solubility Information Soluble in Dichloromethane,Chloroforn,Ether and Methanol
Formula Weight 103.17
Percent Purity 95%
Chemical Name or Material 3-Amino-2,2-dimethyl-1-propanol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.