missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-1H-1,2,4-triazole, 96%

Product Code. 11412667
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11412667 1000 g 1000g
11422667 50 g 50g
11432667 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11412667 Supplier Thermo Scientific Alfa Aesar Supplier No. A14871.0B
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 61-82-5 | C2H4N4 | 84.08 g/mol

3-Amino-1,2,4-triazole is an inhibitor of mitochondrial and chloroplast function. Commercial grade 3- amino-1,2,4-triazole (which generally contains catalase anti-inhibitory impurities) is used as a cotton defoliant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Amino-1,2,4-triazole is an inhibitor of mitochondrial and chloroplast function. Commercial grade 3- amino-1,2,4-triazole (which generally contains catalase anti-inhibitory impurities) is used as a cotton defoliant.

Solubility
It is soluble in water, methanol, ethanol and chloroform, and sparingly soluble in ethyl acetate. It is insoluble in ether and acetone.

Notes
Store desiccated at -20°C. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: -20°C. Keep away from Strong oxidizing agents, Strong acids, Copper, Iron and iron salts.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 61-82-5
Molecular Formula C2H4N4
Molecular Weight (g/mol) 84.08
MDL Number MFCD00005230,MFCD00053362
InChI Key KLSJWNVTNUYHDU-UHFFFAOYSA-N
Synonym 3-amino-1,2,4-triazole, amitrole, aminotriazole, 4h-1,2,4-triazol-3-amine, 1h-1,2,4-triazol-3-amine, amitrol, amerol, amizol, emisol, vorox
PubChem CID 1639
ChEBI CHEBI:40036
IUPAC Name 1H-1,2,4-triazol-5-amine
SMILES NC1=NC=NN1

Specifications

Melting Point 147°C to 153°C
Density 1.138
Assay Percent Range 96%
Odor Odorless
Quantity 1000 g
UN Number UN3077
Beilstein 107687
Merck Index 14,489
Solubility Information It is soluble in water,methanol,ethanol and chloroform,and sparingly soluble in ethyl acetate. It is insoluble in ether and acetone.
Formula Weight 84.08
Percent Purity 96%
Chemical Name or Material 3-Amino-1H-1,2,4-triazole
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.