missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-1H-1,2,4-triazole, 95%

Product Code. p-58888 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
10 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
10g
This item is not returnable. View return policy

Product Code. 10456383

Brand: Thermo Scientific Acros 264570100

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 61-82-5 | C2H4N4 | 84.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 61-82-5
Molecular Formula C2H4N4
Molecular Weight (g/mol) 84.08
MDL Number MFCD00005230,MFCD00053362
InChI Key KLSJWNVTNUYHDU-UHFFFAOYSA-N
Synonym 3-amino-1,2,4-triazole, amitrole, aminotriazole, 4h-1,2,4-triazol-3-amine, 1h-1,2,4-triazol-3-amine, amitrol, amerol, amizol, emisol, vorox
PubChem CID 1639
ChEBI CHEBI:40036
IUPAC Name 1H-1,2,4-triazol-5-amine
SMILES NC1=NC=NN1

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 149°C to 156°C
Color White or Yellow
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Quantity 10 g
Beilstein 26, 137
Merck Index 15, 485
Solubility Information Solubility in water: 280g/L (20°C). Other solubilities: 520g/L (50°C),sparingly soluble in chloroform,dichloromethane,and acetonitrile,slightly soluble in ethylacetate,pract.insol. in diethylether,hydrocarbons,acetone
Formula Weight 84.08
Percent Purity 95%
Physical Form Powder or Flakes
Chemical Name or Material 3-Amino-1H-1, 2, 4-triazole, 95%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.