missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-1H-1,2,4-triazole-5-carboxylic acid hydrate, 98%, Thermo Scientific™

Catalog Number 10370784
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
1266615-63-7, 3641-13-2
C3H4N4O2
128.09
MFCD00010620 MFCD00179325 MFCD00225624
MVRGLMCHDCMPKD-UHFFFAOYSA-N
3-amino-1H-1,2,4-triazole-5-carboxylic acid
NC1=NNC(=N1)C(O)=O
This item is not returnable. View return policy
1266615-63-7, 3641-13-2
C3H4N4O2
128.09
MFCD00010620 MFCD00179325 MFCD00225624
MVRGLMCHDCMPKD-UHFFFAOYSA-N
3-amino-1H-1,2,4-triazole-5-carboxylic acid
NC1=NNC(=N1)C(O)=O
Melting Point 178.0°C to 183.0°C
Color White
Loss on Drying 2% max.
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 100 g
Synonym 5-Amino-2H-1, 2, 4-triazole-3-carboxylic acid hydrate
Solubility Information Solubility in water: .. Other solubilities: insoluble in dil. acid- soluble in aq. alkalis
Formula Weight 128.09
Percent Purity 98%
Physical Form Crystalline Powder and/or Chunks
Chemical Name or Material 3-Amino-1H-1, 2, 4-triazole-5-carboxylic acid hydrate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'