missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-1-propanesulfonic acid, 95%

Product Code. 10085853 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10085853 5 g Glass bottle 5g
10774781 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10085853 Supplier Thermo Scientific Acros Supplier No. 104430050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3687-18-1 | C3H9NO3S | 139.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3687-18-1
Molecular Formula C3H9NO3S
Molecular Weight (g/mol) 139.17
MDL Number MFCD00008225
InChI Key SNKZJIOFVMKAOJ-UHFFFAOYSA-N
Synonym homotaurine, 3-amino-1-propanesulfonic acid, tramiprosate, alzhemed, 3-aminopropanesulphonic acid, 3aps, 1-propanesulfonic acid, 3-amino, cerebril, 3-aminopropanesulfonic acid, 3-aminopropane sulfonic acid
PubChem CID 1646
ChEBI CHEBI:1457
IUPAC Name 3-aminopropane-1-sulfonic acid
SMILES NCCCS(O)(=O)=O

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point >270°C
Color Blue to White
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Linear Formula H2N(CH2)3SO3H
Quantity 5 g
Merck Index 15, 9728
Formula Weight 139.17
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material 3-Amino-1-propanesulfonic acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.