missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-1-phenylbutane, 98%, Thermo Scientific Chemicals

Catalog Number 11433088
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy
22374-89-6
C10H15N
149.237
MFCD00008090
WECUIGDEWBNQJJ-UHFFFAOYSA-N
1-methyl-3-phenylpropylamine, 3-amino-1-phenylbutane, 2-amino-4-phenylbutane, 1-phenyl-3-aminobutane, 4-phenyl-2-aminobutane, 4-phenyl-2-butylamine, 1-phenyl-3-amino-butan, benzenepropanamine, .alpha.-methyl, alpha-methylbenzenepropanamine, propylamine, 1-methyl-3-phenyl
31160
4-phenylbutan-2-amine
CC(CCC1=CC=CC=C1)N
This item is not returnable. View return policy
22374-89-6
C10H15N
149.237
MFCD00008090
WECUIGDEWBNQJJ-UHFFFAOYSA-N
1-methyl-3-phenylpropylamine, 3-amino-1-phenylbutane, 2-amino-4-phenylbutane, 1-phenyl-3-aminobutane, 4-phenyl-2-aminobutane, 4-phenyl-2-butylamine, 1-phenyl-3-amino-butan, benzenepropanamine, .alpha.-methyl, alpha-methylbenzenepropanamine, propylamine, 1-methyl-3-phenyl
31160
4-phenylbutan-2-amine
CC(CCC1=CC=CC=C1)N

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a chemical intermediate. 1-Methyl-3-phenylpropylamine is used in the synthesis of N-substituted derivatives of (1-methyl-3- phenylpropyl)amine.

Solubility
Slightly soluble in water. Soluble in chloroform and ethyl acetate.

Notes
Air Sensitive. Store away from air, oxidizing agents and carbon dioxide. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
TRUSTED_SUSTAINABILITY
Melting Point -50°C
Density 0.928
Boiling Point 220°C to 222°C
Flash Point 97°C (206°F)
Odor Amine-like
Refractive Index 1.514
Quantity 100 g
UN Number UN2922
Beilstein 2413110
Sensitivity Air Sensitive
Solubility Information Slightly soluble in water. Soluble in chloroform and ethyl acetate.
Formula Weight 149.24
Percent Purity 98%
Chemical Name or Material 3-Amino-1-phenylbutane
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'