missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-1-bromoisoquinoline, 97+%

Product Code. 11385349
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11385349 5 g 5g
11375349 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11385349 Supplier Thermo Scientific Alfa Aesar Supplier No. H50368.06
 Restricted Item
Estimated Shipment: 18-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 13130-79-5 | C9H7BrN2 | 223.073 g/mol

3-Amino-1-bromoisoquinoline is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Amino-1-bromoisoquinoline is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Air sensitive. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13130-79-5
Molecular Formula C9H7BrN2
Molecular Weight (g/mol) 223.073
MDL Number MFCD00102193
InChI Key PSQUIUNIVDKHJK-UHFFFAOYSA-N
Synonym 3-amino-1-bromoisoquinoline, 1-bromo-3-isoquinolinamine, 1-bromoisoquinolin-3-ylamine, 1-bromo-3-isoquinolylamine, 3-amino-1-bromo-isoquinoline, 1-bromoisoquinoline-3-amine, acmc-1by0f, 1-bromo-3-aminoisoquinoline, 1-bromanylisoquinolin-3-amine, 3-isoquinolinamine, 1-bromo
PubChem CID 289845
IUPAC Name 1-bromoisoquinolin-3-amine
SMILES C1=CC=C2C(=C1)C=C(N=C2Br)N

Specifications

Quantity 5 g
Sensitivity Air Sensitive
Solubility Information Slightly soluble in water.
Formula Weight 223.07
Percent Purity ≥97%
Chemical Name or Material 3-Amino-1-bromoisoquinoline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.