missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Beskrivelse
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Tekniske data
Tekniske data
| CAS Min % | 95.0 |
| CAS Max % | 100.0 |
| Melting Point | 263°C to 264°C |
| Color | White to Yellow |
| Elemental Analysis | 64.77 to 71.55% C, 7.55 to 8.35% N |
| Flash Point | 110°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 96% |
| Packaging | Glass bottle |
| Quantity | 5 g |
| Vis mere |
Sikkerhed og håndtering
Produktidentifikator
- 3-Amino-1-adamantanol
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?