missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-3-Amino-1,2-propanediol, 97%

Product Code. 11455307
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11455307 500 g 500g
11435307 25 g 25g
11445307 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11455307 Supplier Thermo Scientific Alfa Aesar Supplier No. B22672.36
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 616-30-8 | C3H9NO2 | 91.11 g/mol

3-Amino-1,2-propanediol is used in the preparation of iohexol, which is a X-CT non-ionic contrast medium. It acts as a starting material in the production of specialty materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Amino-1,2-propanediol is used in the preparation of iohexol, which is a X-CT non-ionic contrast medium. It acts as a starting material in the production of specialty materials.

Solubility
Miscible with water.

Notes
Hygroscopic. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 616-30-8
Molecular Formula C3H9NO2
Molecular Weight (g/mol) 91.11
MDL Number MFCD00008140
InChI Key KQIGMPWTAHJUMN-UHFFFAOYSA-N
Synonym 3-amino-1,2-propanediol, 2,3-dihydroxypropylamine, 1,2-propanediol, 3-amino, 1-amino-2,3-propanediol, 1-aminoglycerol, 2,3-propandiol-1-amine, 1-aminopropanediol, 3-aminopropan-1,2-diol, 3-amino-1,2-proopanediol, +/--3-amino-1,2-propanediol
PubChem CID 73561
IUPAC Name 3-aminopropane-1,2-diol
SMILES C(C(CO)O)N

Specifications

Density 1.186
Boiling Point 264°C to 266°C (739 mmHg)
Flash Point 186°C (366°F)
Refractive Index 1.492
Quantity 500 g
UN Number UN2735
Beilstein 1719121
Sensitivity Hygroscopic
Solubility Information Miscible with water.
Formula Weight 91.11
Percent Purity 97%
Chemical Name or Material (+/-)-3-Amino-1,2-propanediol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.