missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-1,2,4-triazole (Amitrole), TRC

Product Code. 30330225
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30330225 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30330225 Supplier Toronto Research Chemicals Supplier No. TRCA633380100G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.
, 02 950 59 478

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Amitrole
CAS 61-82-5
Molecular Formula C2 H4 N4
Quantity 100 g
Synonym 1H-1,2,4-Triazol-3-amine (9CI), 1H-1,2,4-Triazole, 3-amino- (7CI), s-Triazole, 3-amino- (8CI), (4H-1,2,4-Triazol-3-yl)amine, 1,2,4-Triazol-5-amine, 1H-1,2,4-Triazol-3-ylamine, 1H-1,2,4-Triazolamine, 2,3,5,6-Tetraazabicyclo[2.1.1]hex-1-ene, 2-Amino-1,3,4-triazole, 3,A-T, 3-Amino-1,2,4-triazole, 3-Amino-1H-1,2,4-triazole, 3-Amino-2H-1,2,4-triazole, 3-Amino-s-triazole, 3-Aminotriazole, 4H-3-Amino-1,2,4-triazole, 5-Amino-1,2,4-triazole, 5-Amino-1H-1,2,4-triazole, A 0432, AT, ATA, ATA (amine), Amerol, Aminotriazole (plant regulator), Amitrol, Amitrol 90, Amitrol T, Amitrole, Amizol, Azaplant, Azolan, Cytrol, Cytrole, ENT 25445, Herbidal total, Herbizole, Maxata, NSC 34809, NSC 7243, TMG, Weedazol
InChI Formula InChI=1S/C2H4N4/c3-2-4-1-5-6-2/h1H,(H3,3,4,5,6)
SMILES Nc1nc[nH]n1
IUPAC Name 1H-1,2,4-triazol-3-amine
Molecular Weight (g/mol) 84.08
Formula Weight 84.0436
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.