missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-1,2,4-triazole-5-carboxylic acid hydrate, 98%, Thermo Scientific™

Product Code. 11487686
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11487686 50 g 50g
11497686 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11487686 Supplier Thermo Scientific Alfa Aesar Supplier No. A14690.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
3-Amino-1,2,4-triazole-5-carboxylic acid hydrate (AmTAZAc) has been used in the synthesis of new three-dimensional metal-organic framework, [ZnF(AmTAZ)] solvents.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 304655-78-5
Molecular Formula C3H4N4O2
Molecular Weight (g/mol) 128.09
MDL Number MFCD00010620 MFCD00179325 MFCD00225624
InChI Key MVRGLMCHDCMPKD-UHFFFAOYSA-N
Synonym 3-amino-1,2,4-triazole-5-carboxylic acid hydrate, 1h-1,2,4-triazole-3-carboxylicacid, 5-amino-, hydrate 1:1, 3-amino-5-carboxy-1h-1,2,4-triazole hydrate, 5-amino-2h-1,2,4-triazole-3-carboxylic acid hydrate, 3-amino-1,2,4-triazole-5-carboxylic acid hemihydrate, 3-amino-1h-1,2,4-triazole-5-carboxylic acid hydrate, 3-amino-1h-1,2,4-triazole-5-carboxylic acid, oxamethane, 5-amino-1 h-1,2,4-triazole-3-carboxylic acid, 5-amino-4 h-1,2,4 triazole-3-carboxylic acid
PubChem CID 16211818
IUPAC Name 3-amino-1H-1,2,4-triazole-5-carboxylic acid;hydrate
SMILES NC1=NNC(=N1)C(O)=O

Specifications

Melting Point ∼182°C (decomposition)
Quantity 50 g
Beilstein 124734
Solubility Information Slightly soluble in water.
Formula Weight 128.09 (Anhydrous)
Percent Purity 98%
Chemical Name or Material 3-Amino-1,2,4-triazole-5-carboxylic acid hydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.