missing translation for 'onlineSavingsMsg'
Learn More

3-Allylsalicylaldehyde, 97%

Product Code. p-7035748
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15480328 5 g 5g
15490328 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15480328 Supplier Thermo Scientific Alfa Aesar Supplier No. H53475.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 24019-66-7 | C10H10O2 | 162.188 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
3-Allylsalicylaldehyde is a synthetic intermediate for the synthesis of 3-aminoflavones. Flavonoids have been studied for their antiproliferative activity and in vitro cytotoxicity. It is used in the preparation of oxazole derivatives and also imidazole scaffold-based 2-substituted benzofurans for application against human tumor cell lines. It is a reagent in the synthesis of antioxidant and antimicrobial novel chalcones.

Solubility
Soluble in chloroform.

Notes
Air sensitive. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24019-66-7
Molecular Formula C10H10O2
Molecular Weight (g/mol) 162.188
MDL Number MFCD00037364
InChI Key INLWEXRRMUMHKB-UHFFFAOYSA-N
Synonym 3-allyl-2-hydroxybenzaldehyde, 3-allylsalicylaldehyde, 3-allyl-2-hydroxy-benzaldehyde, benzaldehyde, 2-hydroxy-3-2-propenyl, salicylaldehyde, 3-allyl, 2-hydroxy-3-prop-2-en-1-yl benzaldehyde, acmc-209wev, tpc-sk001, 2-hydroxy-3-allylbenzaldehyde
PubChem CID 141062
IUPAC Name 2-hydroxy-3-prop-2-enylbenzaldehyde
SMILES C=CCC1=CC=CC(=C1O)C=O

Specifications

Density 1.099
Flash Point 107°C (225°F)
Refractive Index 1.5645
Quantity 5 g
Sensitivity Air Sensitive
Solubility Information Soluble in chloroform.
Formula Weight 162.19
Percent Purity 97%
Chemical Name or Material 3-Allylsalicylaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.