missing translation for 'onlineSavingsMsg'
Learn More

3-Allyl-5,6-dimethyl-2-thioxo-2,3-dihydrothieno[2,3-d]pyrimidin-4(1H)-one, 96%, Thermo Scientific™

Product Code. p-7032815
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15485205 250 mg 250mg
15468935 1 g 1g
15478935 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15485205 Supplier Thermo Scientific Alfa Aesar Supplier No. H34437.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51486-16-9
Molecular Formula C11H12N2OS2
Molecular Weight (g/mol) 252.35
MDL Number MFCD01911592
InChI Key UIDZXXIBHNLIGD-UHFFFAOYSA-N
Synonym 3-allyl-2-mercapto-5,6-dimethyl-3h-thieno 2,3-d pyrimidin-4-one, 3-allyl-5,6-dimethyl-2-thioxo-2,3-dihydrothieno 2,3-d pyrimidin-4 1h-one, 5,6-dimethyl-3-prop-2-en-1-yl-2-sulfanyl-3h,4h-thieno 2,3-d pyrimidin-4-one, 3-allyl-2-mercapto-5,6-dimethylthieno 2,3-d pyrimidin-4 3h-one, thieno 2,3-d pyrimidin-4 1h-one,2,3-dihydro-5,6-dimethyl-3-2-propen-1-yl-2-thioxo, 5,6-dimethyl-3-prop-2-en-1-yl-2-sulfanylidene-1h-thieno 2,3-d pyrimidin-4-one, 5,6-dimethyl-3-prop-2-en-1-yl-2-sulfanylthieno 2,3-d pyrimidin-4-one, 5,6-dimethyl-3-prop-2-enyl-2-thioxo-1,3-dihydrothiopheno 2,3-d pyrimidin-4-one, 3-allyl-2-mercapto-5,6-dimethyl-4-oxo-3,4-dihydrothieno 2,3-d pyrimidine
PubChem CID 722257
IUPAC Name 5,6-dimethyl-3-prop-2-enyl-2-sulfanylidene-1H-thieno[2,3-d]pyrimidin-4-one
SMILES CC1=C(C)C2=C(NC(=S)N(CC=C)C2=O)S1

Specifications

Melting Point 202°C to 203°C
Quantity 250 mg
Formula Weight 252.36
Percent Purity 96%
Chemical Name or Material 3-Allyl-5,6-dimethyl-2-thioxo-2,3-dihydrothieno[2,3-d]pyrimidin-4(1H)-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.