missing translation for 'onlineSavingsMsg'
Learn More

3-(Acryloyloxy)propyltrimethoxysilane, 94%, stab. with 100ppm BHT

Product Code. 11319155
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11319155 25 g 25g
11309155 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11319155 Supplier Thermo Scientific Alfa Aesar Supplier No. L16400.14
  • ONLINE EXCLUSIVE PRICE
    £144.84
    £116.65 / 25g
    Save £28.19
    19% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 4369-14-6 | C9H18O5Si | 234.32 g/mol

3-(Acryloyloxy)propyltrimethoxysilane is used to enhance the thermal conductivity of silicone rubber filled with hybrid fillers. It is involved in the synthesis of vinyl and acryalte modified silica nanoparticles by using vinyltrimethoxysilane. Further, it is used as a silane coupling agent involved in the modification of fluorescein isothiocyanate-mesoporous silica nanoparticles with carbon-carbon double bonds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-(Acryloyloxy)propyltrimethoxysilane is used to enhance the thermal conductivity of silicone rubber filled with hybrid fillers. It is involved in the synthesis of vinyl and acryalte modified silica nanoparticles by using vinyltrimethoxysilane. Further, it is used as a silane coupling agent involved in the modification of fluorescein isothiocyanate-mesoporous silica nanoparticles with carbon-carbon double bonds.

Solubility
Miscible with organic solvents.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents, acids and bases. Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4369-14-6
Molecular Formula C9H18O5Si
Molecular Weight (g/mol) 234.32
MDL Number MFCD00054803
InChI Key KBQVDAIIQCXKPI-UHFFFAOYSA-N
Synonym 3-trimethoxysilyl propyl acrylate, 3-acryloyloxy propyltrimethoxysilane, 3-acryloxypropyltrimethoxysilane, 2-propenoic acid, 3-trimethoxysilyl propyl ester, 3-acryloxypropyl trimethoxysilane, 3-trimethoxysilyl propyl 2-propenoate, 3-acryloyloxypropyltrimethoxysilane, 3-acryloxypropyltrimethoxysilane, stab. with 100ppm bht, acmc-1aoz6, acrylic acid, 3-trimethoxysilyl propyl ester
PubChem CID 151204
IUPAC Name 3-trimethoxysilylpropyl prop-2-enoate
SMILES CO[Si](CCCOC(=O)C=C)(OC)OC

Specifications

Boiling Point 95°C to 100°C (3mmHg)
Quantity 25 g
Flash Point 123°C (253°F)
Refractive Index 1.429
Sensitivity Moisture sensitive
Density 1.059
Linear Formula CH2=CHCO2(CH2)3Si(OCH3)3
UN Number UN1760
Beilstein 2046320
Solubility Information Miscible with organic solvents.
Formula Weight 234.33
Percent Purity 94%
Chemical Name or Material 3-(Acryloyloxy)propyltrimethoxysilane, stabilized with 100ppm BHT
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 3-(Acryloyloxy)propyltrimethoxysilane
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Skin sensitiser Category 1
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H332-Harmful if inhaled.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: 2-Propenoic acid, 3-(trimethoxysilyl)propylester

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.