missing translation for 'onlineSavingsMsg'
Learn More

3-Acetylthiophene, 98%

Product Code. 11408943
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11408943 10 g 10g
11418943 50 g 50g
11428943 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11408943 Supplier Thermo Scientific Alfa Aesar Supplier No. A11078.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1468-83-3 | C6H6OS | 126.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1468-83-3
Molecular Formula C6H6OS
Molecular Weight (g/mol) 126.17
MDL Number MFCD00005468
InChI Key RNIDWJDZNNVFDY-UHFFFAOYSA-N
Synonym 3-acetylthiophene, methyl 3-thienyl ketone, 1-thiophen-3-yl ethan-1-one, 1-thiophen-3-yl ethanone, 1-3-thienyl ethanone, ethanone, 1-3-thienyl, ketone, methyl 3-thienyl, 3-acetyl thiophene, 1-thien-3-ylethanone, methyl-3-thienyl ketone
PubChem CID 15116
IUPAC Name 1-thiophen-3-ylethanone
SMILES CC(=O)C1=CSC=C1

Specifications

Melting Point 58°C to 61°C
Boiling Point 208°C to 210°C
Quantity 10 g
UN Number UN2811
Beilstein 107241
Sensitivity Light sensitive
Formula Weight 126.18
Percent Purity 98%
Chemical Name or Material 3-Acetylthiophene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.