missing translation for 'onlineSavingsMsg'
Learn More

3-Acetylpyridine, TRC

Product Code. 30040625
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30040625 10 g 10g
18031830 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30040625 Supplier Toronto Research Chemicals Supplier No. TRCA18760010G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 350-03-8
Molecular Formula C7 H7 N O
Molecular Weight (g/mol) 121.14
Synonym 1-(Pyridin-3-yl)ethan-1-one, 3-Acetylpyridine
IUPAC Name 1-pyridin-3-ylethanone
SMILES CC(=O)c1cccnc1

Specifications

Quantity 10 g
InChI Formula InChI=1S/C7H7NO/c1-6(9)7-3-2-4-8-5-7/h2-5H,1H3
Formula Weight 121.0528
Chemical Name or Material 1-(pyridin-3-yl)ethan-1-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.