missing translation for 'onlineSavingsMsg'
Learn More

3-Acetylpyridine adenine dinucleotide, 90%

Product Code. 15555986 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15555986 250 mg 250mg
15545986 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15555986 Supplier Thermo Scientific Acros Supplier No. 457682500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 86-08-8 | C22H28N6O14P2 | 662.44 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C22H28N6O14P2
Molecular Weight (g/mol) 662.44
MDL Number MFCD00078882
InChI Key KPVQNXLUPNWQHM-RBEMOOQDSA-N
Synonym 3-acetylpyridine adenine dinucleotide, acpyad, 3-acetyl nad, 3-acetylpyridine-dpn, 3-acetylpyridine ad, dpn 3-acetylpyridine analog, 3-acetylpyridine analog of dpn, acetylpyridine adenine dinucleotide, acetylpyridine-adenine dinucleotide, 3-acetylpyridineadenine dinucleotide
PubChem CID 123926
SMILES CC(=O)C1=CC=C[N+](=C1)[C@@H]1O[C@H](COP([O-])(=O)OP(O)(=O)OC[C@H]2O[C@H]([C@H](O)[C@@H]2O)N2C=NC3=C(N)N=CN=C23)[C@@H](O)[C@H]1O

Specifications

Infrared Spectrum Authentic
Assay Percent Range 89% min. (HPLC)
Quantity 250 mg
IUPAC Name 3-acetyl-1-[(2R,3R,4S,5R)-5-{[({[(2R,3S,4R,5R)-5-(6-amino-9H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy}(hydroxy)phosphoryl phosphonato)oxy]methyl}-3,4-dihydroxyoxolan-2-yl]-1λ⁵-pyridin-1-ylium
Formula Weight 662.45
Percent Purity 90%
Chemical Name or Material 3-Acetylpyridine adenine dinucleotide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.