missing translation for 'onlineSavingsMsg'
Learn More

3-Acetylphenyl isothiocyanate, 97%

Product Code. 11378245
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11378245 5 g 5g
11368245 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11378245 Supplier Thermo Scientific Alfa Aesar Supplier No. L10134.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 3125-71-1 | C9H7NOS | 177.221 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3125-71-1
Molecular Formula C9H7NOS
Molecular Weight (g/mol) 177.221
MDL Number MFCD00041081
InChI Key RGANVWCPAAIVNN-UHFFFAOYSA-N
Synonym 3-acetylphenyl isothiocyanate, 1-3-isothiocyanatophenyl ethanone, 3-acetylphenylisothiocyanate, 1-3-isothiocyanato-phenyl-ethanone, 3-acetylbenzenisothiocyanate, acmc-20anak, 3'-isothiocyanatoacetophenone, 3-methylcarbonylphenyl isothiocyanate, 1-3-isothiocyanatophenyl ethanone #, ethanone,1-3-isothiocyanatophenyl
PubChem CID 520515
IUPAC Name 1-(3-isothiocyanatophenyl)ethanone
SMILES CC(=O)C1=CC(=CC=C1)N=C=S

Specifications

Melting Point ∼24°C
Boiling Point 135°C to 136°C (2 mmHg)
Flash Point >110°C (230°F)
Odor Irritating
Refractive Index 1.648
Quantity 5 g
UN Number UN2923
Beilstein 2803021
Sensitivity Moisture sensitive
Formula Weight 177.23
Percent Purity 97%
Chemical Name or Material 3-Acetylphenyl isothiocyanate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.