missing translation for 'onlineSavingsMsg'
Learn More

3-Acetylphenyl isocyanate, 97%, Thermo Scientific™

Product Code. 10083740 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Glass bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10083740 10 g Glass bottle 10g
10209513 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10083740 Supplier Thermo Scientific Acros Supplier No. 314100100
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 23138-64-9
Molecular Formula C9H7NO2
Molecular Weight (g/mol) 161.16
MDL Number MFCD00013866
InChI Key JASTZOAHAWBRLM-UHFFFAOYSA-N
Synonym 1-3-isocyanatophenyl ethanone, 3-acetylphenyl isocyanate, 3-acetylphenylisocyanate, 3-isocyanatoacetophenone, ethanone,1-3-isocyanatophenyl, ethanone, 1-3-isocyanatophenyl, 3-acetylbenzenisocyanate, 3'-isocyanatoacetophenone, acmc-1cjm8, 3'-isocyanato-acetophenone
PubChem CID 312718
IUPAC Name 1-(3-isocyanatophenyl)ethanone
SMILES CC(=O)C1=CC=CC(=C1)N=C=O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 33°C to 34°C
Color Yellow to Brown
Density 1.1700g/mL
Boiling Point 155°C (4.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula CH3COC6H4NCO
Refractive Index 1.5610 to 1.563
Quantity 10 g
Specific Gravity 1.17
Formula Weight 161.16
Percent Purity 97%
Physical Form Liquid After Melting
Chemical Name or Material 3-Acetylphenyl isocyanate, 97%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.