missing translation for 'onlineSavingsMsg'
Learn More

3-Acetylphenyl Ethyl(methyl)carbamate, Mikromol™

Product Code. 17983796
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
This item is not returnable. View return policy

Product Code. 17983796

Brand: Mikromol MM1497.050025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

3-Acetylphenyl Ethyl(methyl)carbamate
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-Acetylphenyl Ethyl(methyl)carbamate
CAS 855300-09-3
Analyte or Component Names 3-Acetylphenyl Ethyl(methyl)carbamate
Shipping Condition Room Temperature
Molecular Formula C12 H15 N O3
Certifications/Compliance ISO 17025
Synonym 3-Acetylphenyl Ethyl(methyl)carbamate, Des-[3-(1-Dimethylamino)ethyl] 3-Acetylrivastigmine, Rivastigmine Related Compound D (USP), Rivastigmine Tartrate Imp. C (EP), Rivastigmine Hydrogen Tartrate Imp. C (EP), Rivastigmine Imp. C (EP), Rivastigmine USP Related Compound D, Rivastigmine USP RC D, Rivastigmine Related Compound D, Rivastigmine Hydrogen Tartrate Impurity C, Rivastigmine Impurity C
InChI Formula InChI=1S/C12H15NO3/c1-4-13(3)12(15)16-11-7-5-6-10(8-11)9(2)14/h5-8H,4H2,1-3H3
SMILES CCN(C)C(=O)Oc1cccc(c1)C(=O)C
IUPAC Name (3-acetylphenyl) N-ethyl-N-methylcarbamate
Molecular Weight (g/mol) 221.25
Quantity 25 mg
Formula Weight 221.1052 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.