missing translation for 'onlineSavingsMsg'
Learn More

3-Acetylindole, TRC

Product Code. 30318476
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30318476 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30318476 Supplier Toronto Research Chemicals Supplier No. TRCA19222810MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-Acetylindole
CAS 703-80-0
Molecular Formula C10H9NO
Quantity 10 mg
Synonym 1-(1H-Indol-3-yl)ethanone, 1-(1H-Indol-3-yl)ethan-1-one, 3-Acetyl-1H-indole, 3-Acetylindole, 3-Indolyl Methyl Ketone, NSC 47180, NSC 58084
InChI Formula InChI=1S/C10H9NO/c1-7(12)9-6-11-10-5-3-2-4-8(9)10/h2-6,11H,1H3
SMILES CC(=O)c1c[nH]c2ccccc12
IUPAC Name 1-(1H-indol-3-yl)ethanone
Molecular Weight (g/mol) 159.18
Formula Weight 159.0684
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.