missing translation for 'onlineSavingsMsg'
Learn More

3-Acetylbenzonitrile, 97%, Thermo Scientific™

Product Code. 10750781 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass Bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10750781 5 g Glass Bottle 5g
10261383 1 g Glass Bottle 1g
10311144 25 g Glass Bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10750781 Supplier Thermo Scientific Acros Supplier No. 307840050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6136-68-1
Molecular Formula C9H7NO
Molecular Weight (g/mol) 145.16
MDL Number MFCD00001806
InChI Key SBCFGFDAZCTSRH-UHFFFAOYSA-N
Synonym m-cyanoacetophenone, 3'-cyanoacetophenone, benzonitrile, 3-acetyl, 3-cyanoacetophenone, 3-acetyl-benzonitrile, unii-tsj72034p9, 3-acetylbenzenecarbonitrile, m-acetylbenzonitrile, 3-acetyl benzonitrile, pubchem12649
PubChem CID 80222
IUPAC Name 3-acetylbenzonitrile
SMILES CC(=O)C1=CC=CC(=C1)C#N

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 96.0°C to 100.0°C
Color Brown to Orange-Yellow
Boiling Point 120.0°C (5.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass Bottle
Linear Formula CH3COC6H4CN
Quantity 5 g
Formula Weight 145.16
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 3-Acetylbenzonitrile
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.