missing translation for 'onlineSavingsMsg'
Learn More

3-Acetylbenzonitrile, 97+%

Product Code. 11419812
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
5 g
25 g
100 g
missing translation for 'unitSize'
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11419812 5 g 5g
11429812 25 g 25g
11439812 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 11419812 missing translation for 'mfr' Thermo Scientific Alfa Aesar missing translation for 'supplierNo' A10841.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6136-68-1 | C9H7NO | 145.16 g/mol

Preparation of (q2-2-Acetyl-4-cyanophenyl)tetra- carbonylmanganese by Reaction of 3-Acetylbenzonitrile with Benzylpentacarbonylmanganese. Four aromatic amidoximes were synthesized by reacting hydroxylamine with 1,3 -dicyanobenzene, 1,4- dicyanobenzene, 3-acetylbenzonitrile, and 4-acetylbenzonitrile. Darzens condensation of 3-acetylbenzonitrile provided aldehyde.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Preparation of (q2-2-Acetyl-4-cyanophenyl)tetra- carbonylmanganese by Reaction of 3-Acetylbenzonitrile with Benzylpentacarbonylmanganese. Four aromatic amidoximes were synthesized by reacting hydroxylamine with 1,3 -dicyanobenzene, 1,4- dicyanobenzene, 3-acetylbenzonitrile, and 4-acetylbenzonitrile. Darzens condensation of 3-acetylbenzonitrile provided aldehyde.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6136-68-1
Molecular Formula C9H7NO
Molecular Weight (g/mol) 145.16
MDL Number MFCD00001806
InChI Key SBCFGFDAZCTSRH-UHFFFAOYSA-N
Synonym m-cyanoacetophenone, 3'-cyanoacetophenone, benzonitrile, 3-acetyl, 3-cyanoacetophenone, 3-acetyl-benzonitrile, unii-tsj72034p9, 3-acetylbenzenecarbonitrile, m-acetylbenzonitrile, 3-acetyl benzonitrile, pubchem12649
PubChem CID 80222
IUPAC Name 3-acetylbenzonitrile
SMILES CC(=O)C1=CC=CC(=C1)C#N

Specifications

Melting Point 97°C to 101°C
Boiling Point 120°C (5 mmHg)
Quantity 5 g
UN Number UN3439
Beilstein 2079629
Solubility Information Insoluble in water.
Formula Weight 145.16
Percent Purity ≥97%
Chemical Name or Material 3-Acetylbenzonitrile

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.