missing translation for 'onlineSavingsMsg'
Learn More

3-Acetylbenzoic acid, 98%

Product Code. 11491637
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11491637 1 g 1g
11401647 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11491637 Supplier Thermo Scientific Alfa Aesar Supplier No. B25616.03
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 586-42-5 | C9H8O3 | 164.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 586-42-5
Molecular Formula C9H8O3
Molecular Weight (g/mol) 164.16
MDL Number MFCD00045847
InChI Key CHZPJUSFUDUEMZ-UHFFFAOYSA-N
Synonym m-acetylbenzoic acid, benzoic acid, 3-acetyl, 3-acetylbenzoicacid, 3-acetophenonecarboxylic acid, acetophenone-3-carboxylic acid, 3-acetyl benzoic acid, 3-acetyl-benzoic acid, hypnone-m-carboxylic acid, 3-1-oxoethyl benzoic acid, 3-acetylbenzoic acid
PubChem CID 220324
IUPAC Name 3-acetylbenzoic acid
SMILES CC(=O)C1=CC(=CC=C1)C(=O)O

Specifications

Melting Point 162°C to 166°C
Odor Odorless
Quantity 1 g
Formula Weight 164.16
Percent Purity 98%
Chemical Name or Material 3-Acetylbenzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.