missing translation for 'onlineSavingsMsg'
Learn More

3-Acetylbenzeneboronic acid, 97%

Product Code. 11475477
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11475477 5 g 5g
11465477 1 g 1g
11485477 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11475477 Supplier Thermo Scientific Alfa Aesar Supplier No. B23478.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 204841-19-0 | C8H9BO3 | 163.967 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 204841-19-0
Molecular Formula C8H9BO3
Molecular Weight (g/mol) 163.967
MDL Number MFCD01074678
InChI Key SJGGDZCTGBKBCK-UHFFFAOYSA-N
Synonym 3-acetylphenyl boronic acid, 3-acetylbenzeneboronic acid, 3-acetylphenylboronicacid, m-acetylphenylboronic acid, 3'-boronoacetophenone, boronic acid, 3-acetylphenyl, 1-3-dihydroxyboranyl phenyl ethan-1-one, pubchem2751, acmc-209faq, 3-acetylphenyboronic acid
PubChem CID 2734310
ChEBI CHEBI:86552
IUPAC Name (3-acetylphenyl)boronic acid
SMILES B(C1=CC(=CC=C1)C(=O)C)(O)O

Spécification

Melting Point ∼200°C to 205°C
Quantity 5 g
Beilstein 8255673
Formula Weight 163.97
Percent Purity 97%
Chemical Name or Material 3-Acetylbenzeneboronic acid

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.