missing translation for 'onlineSavingsMsg'
Learn More

3-Acetyl-4-methylpyridine, 96%

Product Code. 15456197
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15456197 5 g 5g
15446197 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15456197 Supplier Thermo Scientific Alfa Aesar Supplier No. H33380.06
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 51227-30-6 | C8H9NO | 135.166 g/mol

It is used as pharmaceutical intermediate. Benzoylthiophenes are allosteric enhancers (AE) of agonist activity at the A1 adenosine receptor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as pharmaceutical intermediate. Benzoylthiophenes are allosteric enhancers (AE) of agonist activity at the A1 adenosine receptor.

Solubility
Sparingly soluble in water.(0.26 g/L) (25°C),

Notes
Store in cool dry place. Ensure proper ventilation. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51227-30-6
Molecular Formula C8H9NO
Molecular Weight (g/mol) 135.166
MDL Number MFCD08447082
InChI Key LCEINQHGZAFADC-UHFFFAOYSA-N
Synonym 1-4-methylpyridin-3-yl ethanone, 3-acetyl-4-methylpyridine, 1-4-methyl-3-pyridinyl ethanone, 1-4-methyl-3-pyridinyl-ethanone, 1-4-methylpyridin-3-yl ethan-1-one, 1-4-methyl-pyridin-3-yl-ethanone, ethanone, 1-4-methyl-3-pyridinyl, 4-methyl-3-acetylpyridine, 3-acetyl-4-picoline, methyl 4-picolyl ketone
PubChem CID 10942481
IUPAC Name 1-(4-methylpyridin-3-yl)ethanone
SMILES CC1=C(C=NC=C1)C(=O)C

Specifications

Boiling Point 57°C to 58°C (1 mmHg)
Refractive Index 1.5295
Quantity 5 g
Solubility Information Sparingly soluble in water.(0.26g/L) (25°C),
Formula Weight 135.17
Percent Purity 96%
Chemical Name or Material 3-Acetyl-4-methylpyridine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.