missing translation for 'onlineSavingsMsg'
Learn More

3-Acetopropanol (mixture of monomer and dimer), TRC

Product Code. 30325782
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30325782 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30325782 Supplier Toronto Research Chemicals Supplier No. TRCA16402010G
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 5-hydroxypentan-2-one
CAS 1071-73-4
Molecular Formula C5H10O2
Quantity 10 g
Synonym 1-Hydroxy-4-pentanone, 3-Acetyl-1-propanol, γ-Acetopropyl Alcohol, 4-Oxo-1-pentanol, 5-Hydroxy-2-pentanone, 5-Hydroxy-n-pentan-2-one, NSC 19158, NSC 33940, γ-Acetopropanol
InChI Formula InChI=1S/C5H10O2/c1-5(7)3-2-4-6/h6H,2-4H2,1H3
SMILES O=C(CCCO)C
IUPAC Name 5-hydroxypentan-2-one
Molecular Weight (g/mol) 102.13
Formula Weight 102.07
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.