missing translation for 'onlineSavingsMsg'
Learn More

3-Acetamidophenol, 99+%

Product Code. 10162510 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10162510 100 g Glass bottle 100g
10538100 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10162510 Supplier Thermo Scientific Acros Supplier No. 102321000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 621-42-1 | C8H9NO2 | 151.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 621-42-1
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.16
MDL Number MFCD00002263
InChI Key QLNWXBAGRTUKKI-UHFFFAOYSA-N
Synonym 3-acetamidophenol, n-3-hydroxyphenyl acetamide, metacetamol, 3'-hydroxyacetanilide, m-acetamidophenol, 3-hydroxyacetanilide, n-acetyl-m-aminophenol, m-hydroxyacetanilide, 3-acetylamino phenol, metalid
PubChem CID 12124
ChEBI CHEBI:76987
IUPAC Name N-(3-hydroxyphenyl)acetamide
SMILES CC(=O)NC1=CC(=CC=C1)O

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 146°C to 149°C
Color Gray or Tan to White
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula CH3CONHC6H4OH
Quantity 100 g
Beilstein 13, 415
Solubility Information Solubility in water: freely soluble in hot water
Formula Weight 151.16
Percent Purity 99+%
Physical Form Crystalline Powder, Crystals or Needles
Chemical Name or Material 3-Acetamidophenol, 99+%
Show More Show Less
compliance-icons
Product Identifier
  • 3-Acetamidophenol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.