missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-Acetamido-2-nitrobenzeneboronic acid, 96%

Product Code. 15470088
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15470088 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15470088 Supplier Alfa Aesar™ Supplier No. H53348.MD
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Applications
3-Acetamido-2-nitrobenzeneboronic acid is used as pharmaceutical intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Specifications

CAS 78887-38-4
Melting Point 161°C to 162°C
Molecular Formula C8H9BN2O5
MDL Number MFCD06659828
Quantity 250 mg
Synonym 3-acetamido-2-nitro benzeneboronic acid, 3-acetamido-2-nitro benzeneboronicacid, 3-acetamido-2-nitrophenyl boronic acid, 3-acetamido-2-nitrobenzeneboronic acid, boronic acid,b-3-acetylamino-2-nitrophenyl, acmc-209pfs, 3-acetamido-2-nitro-phenyl boronic acid, 3-acetylamino-2-nitrophenyl boronic acid, 3-acetylamino-2-nitrophenylboronic acid
InChI Key XJRVFUIJKNFQBX-UHFFFAOYSA-N
SMILES B(C1=C(C(=CC=C1)NC(=O)C)[N+](=O)[O-])(O)O
IUPAC Name (3-acetamido-2-nitrophenyl)boronic acid
Molecular Weight (g/mol) 223.979
PubChem CID 44119623
Formula Weight 223.98
Percent Purity 96%
Chemical Name or Material 3-Acetamido-2-nitrobenzeneboronic acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.