missing translation for 'onlineSavingsMsg'
Learn More

3-(9H-Carbazol-4-yloxy)propane-1,2-diol, Mikromol™

Product Code. 17943746
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Product Code. Quantity unitSize
17943746 25 mg 25mg
1 options
This item is not returnable. View return policy

Product Code. 17943746

Brand: Mikromol MM0291.140025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

3-(9H-Carbazol-4-yloxy)propane-1,2-diol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-(9H-Carbazol-4-yloxy)-1,2-propanediol
CAS 123119-89-1
Analyte or Component Names 3-(9H-Carbazol-4-yloxy)-1,2-propanediol
Shipping Condition Room Temperature
Molecular Formula C15 H15 N O3
Certifications/Compliance ISO 9001
Synonym 1,2-Propanediol, 3-(9H-carbazol-4-yloxy)-, 3-(9H-Carbazol-4-yloxy)-1,2-propanediol, 4-(2,3-Dihydroxy-1-propoxy)carbazole
InChI Formula InChI=1S/C15H15NO3/c17-8-10(18)9-19-14-7-3-6-13-15(14)11-4-1-2-5-12(11)16-13/h1-7,10,16-18H,8-9H2
SMILES OCC(O)COc1cccc2[nH]c3ccccc3c12
IUPAC Name 3-(9H-carbazol-4-yloxy)propane-1,2-diol
Molecular Weight (g/mol) 257.28
Quantity 25 mg
Formula Weight 257.1052 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.