missing translation for 'onlineSavingsMsg'
Learn More

3,8-Diamino-6-phenylphenanthridine, 98%

Product Code. 11456966
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11456966 5 g 5g
11446966 1 g 1g
11466966 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11456966 Supplier Thermo Scientific Alfa Aesar Supplier No. B24049.06
  • ONLINE EXCLUSIVE PRICE
    £223.38
    £183.65 / 5g
    Save £39.73
    17% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 52009-64-0 | C19H15N3 | 285.35 g/mol

3,8-Diamino-6-phenylphenanthridine is used in the synthesis of rigid polyamides. It has anti-tumor and anti-viral properties.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,8-Diamino-6-phenylphenanthridine is used in the synthesis of rigid polyamides. It has anti-tumor and anti-viral properties.

Solubility
Soluble in chloroform, dichloromethane and methanol.

Notes
Store at -20°C. Keep container tightly closed in a dry and well-ventilated place. Incompatible material is strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52009-64-0
Molecular Formula C19H15N3
Molecular Weight (g/mol) 285.35
MDL Number MFCD00075152
InChI Key CPNAVTYCORRLMH-UHFFFAOYSA-N
Synonym 3,8-diamino-6-phenylphenanthridine, 6-phenylphenathridine-3,8-diamine, 3,8-dapp, 3,8-diamino-6-phenyl-phenanthridine, acmc-1aycj, 6-phenyl-3,8-phenanthridinediamine, 3,8-diamino-6-phenyl phenanthridine, 3,8-phenanthridinediamine,6-phenyl, 3,8-diamino-6-phenylphenanthridine6-phenyl-3,8-phenanthridinediamine,
PubChem CID 104052
IUPAC Name 6-phenylphenanthridine-3,8-diamine
SMILES C1=CC=C(C=C1)C2=C3C=C(C=CC3=C4C=CC(=CC4=N2)N)N

Specifications

Melting Point 196°C to 198°C
Quantity 5 g
Solubility Information Soluble in chloroform,dichloromethane and methanol.
Formula Weight 285.35
Percent Purity 98%
Chemical Name or Material 3,8-Diamino-6-phenylphenanthridine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.