missing translation for 'onlineSavingsMsg'
Learn More

3,7-Dimethyl-3-octanol, 97+%

Product Code. 10599033 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Glass bottle
Unit Size:
100mL
500mL
This item is not returnable. View return policy

Product Code. 10599033

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 78-69-3 | C10H22O | 158.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78-69-3
Molecular Formula C10H22O
Molecular Weight (g/mol) 158.28
MDL Number MFCD00004482
InChI Key DLHQZZUEERVIGQ-UHFFFAOYSA-N
Synonym tetrahydrolinalool, 3,7-dimethyl-3-octanol, 3-octanol, 3,7-dimethyl, linalool tetrahydride, 2,6-dimethyl-6-octanol, 3,7-dimethyloctanol-3, fema no. 3060, 1-3,7-dimethyloctan-3-ol, dsstox_cid_9110, dsstox_rid_78670
PubChem CID 6548
ChEBI CHEBI:84242
IUPAC Name 3,7-dimethyloctan-3-ol
SMILES CCC(C)(CCCC(C)C)O

Specifications

CAS Min % 97.0
CAS Max % 100.0
Color Colorless
Density 0.8280g/mL
Boiling Point 193.0°C
Flash Point 76°C
Infrared Spectrum Authentic
Assay Percent Range 97% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)2CH(CH2)3C(CH3)(OH)CH2CH3
Refractive Index 1.4310 to 1.4350
Quantity 100 mL
Specific Gravity 0.828
Solubility Information Solubility in water: .. Other solubilities: soluble in alcohol
Formula Weight 158.28
Percent Purity 97+%
Physical Form Liquid
Chemical Name or Material 3, 7-Dimethyl-3-octanol
Show More Show Less
compliance-icons
Product Identifier
  • 3,7-Dimethyl-3-octanol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.