missing translation for 'onlineSavingsMsg'
Learn More

3,7-Dimethyl-1-octanol, 95%

Product Code. 10300254 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
50 mL
250 mL
1 L
Packaging:
Glass bottle
Unit Size:
1 L
250mL
50mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10300254 1 L Glass bottle 1 L
10648624 50 mL Glass bottle 50mL
10269193 250 mL Glass bottle 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10300254 Supplier Thermo Scientific Supplier No. 392620010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 106-21-8 | C10H22O | 158.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106-21-8
Molecular Formula C10H22O
Molecular Weight (g/mol) 158.28
MDL Number MFCD00002936
InChI Key PRNCMAKCNVRZFX-UHFFFAOYSA-N
Synonym 3,7-dimethyl-1-octanol, tetrahydrogeraniol, dimethyloctanol, dihydrocitronellol, pelargol, perhydrogeraniol, geraniol tetrahydride, citronellol, dihydro, 1-octanol, 3,7-dimethyl, geraniol, perhydro
PubChem CID 7792
IUPAC Name 3,7-dimethyloctan-1-ol
SMILES CC(C)CCCC(C)CCO

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color Colorless
Density 0.8280g/mL
Boiling Point 118°C (15.0 mmHg)
Flash Point 97°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)2CH(CH2)3CH(CH3)(CH2)2OH
Refractive Index 1.4340 to 1.4360 (20°C, 589nm)
Quantity 1 L
Specific Gravity 0.828
Formula Weight 158.28
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 3, 7-Dimethyl-1-octanol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 3,7-Dimethyl-1-octanol
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H411-Toxic to aquatic life with long lasting effects.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.