missing translation for 'onlineSavingsMsg'
Learn More

3,7-Dihydro-4H-imidazo[4,5-d]-1,2,3-triazin-4-one Monohydrate (2-Azahypoxanthine Monohydrate), Mikromol™

Product Code. 17993646
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
This item is not returnable. View return policy

Product Code. 17993646

Brand: Mikromol MM1289.05

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

3,7-Dihydro-4H-imidazo[4,5-d]-1,2,3-triazin-4-one Monohydrate (2-Azahypoxanthine Monohydrate)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Azahypoxanthine Monohydrate
CAS 3-3-7151
Analyte or Component Names 2-Azahypoxanthine Monohydrate
Shipping Condition Room Temperature
Molecular Formula C4 H3 N5 O . H2 O
Certifications/Compliance ISO/IEC 17025
Synonym Dacarbazine Imp. A (EP), Temozolomide Imp. E (EP), Dacarbazine USP Related Compound B, Dacarbazine USP RC B, 3,7-Dihydro-4H-imidazo[4,5-d]-1,2,3-triazin-4-one Monohydrate, 2-Azahypoxanthine Monohydrate, Dacarbazine Related Compound B as Monohydrate, Dacarbazine Impurity A as Monohydrate, Temozolomide Impurity E as Monohydrate
InChI Formula InChI=1S/C4H3N5O.H2O/c10-4-2-3(6-1-5-2)7-9-8-4;/h1H,(H2,5,6,7,8,10);1H2
SMILES O.O=C1NN=Nc2[nH]cnc12
IUPAC Name 3,7-dihydroimidazo[4,5-d]triazin-4-one;hydrate
Molecular Weight (g/mol) 155.11
Quantity 100 mg
Formula Weight 155.0443 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.