missing translation for 'onlineSavingsMsg'
Learn More

3,7-Dihydro-3-methyl-1,7-bis(5-oxohexyl)-1H-purine-2,6-dione, TRC

Product Code. 30384071
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30384071 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30384071 Supplier Toronto Research Chemicals Supplier No. TRCD44746825MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1,7-Bis-(5-oxohexyl)-3-methylxanthine
CAS 200556-62-3
Molecular Formula C18 H26 N4 O4
Quantity 25 mg
Synonym 1,7-Bis-(5-oxohexyl)-3-methylxanthine
InChI Formula InChI=1S/C18H26N4O4/c1-13(23)8-4-6-10-21-12-19-16-15(21)17(25)22(18(26)20(16)3)11-7-5-9-14(2)24/h12H,4-11H2,1-3H3
SMILES CN1C(=O)N(CCCCC(=O)C)C(=O)c2c1ncn2CCCCC(=O)C
IUPAC Name 3-methyl-1,7-bis(5-oxohexyl)purine-2,6-dione
Molecular Weight (g/mol) 362.42
Formula Weight 362.1954
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.