missing translation for 'onlineSavingsMsg'
Learn More

3-[(6-Methylpyrazin-2-yl)oxy]aniline, 97%, Thermo Scientific™

£101.00 - £125.00

Chemical Identifiers

CAS 915707-61-8
Molecular Formula C11H11N3O
Molecular Weight (g/mol) 201.229
MDL Number MFCD09702406
InChI Key ZFDWDKWQAXTCSU-UHFFFAOYSA-N
PubChem CID 24229682
IUPAC Name 3-(6-methylpyrazin-2-yl)oxyaniline
SMILES CC1=CN=CC(=N1)OC2=CC=CC(=C2)N
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10481884
View Documents Promotion Details
Thermo Scientific Maybridge
CC62614CB
250 mg Amber Glass Bottle
£101.00
250mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
10718835
View Documents Promotion Details
Thermo Scientific Maybridge
CC62614DA
1 g Amber Glass Bottle
£125.00
1g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

915707-61-8
201.229
ZFDWDKWQAXTCSU-UHFFFAOYSA-N
3-(6-methylpyrazin-2-yl)oxyaniline
C11H11N3O
MFCD09702406
24229682
CC1=CN=CC(=N1)OC2=CC=CC(=C2)N

Specifications

915707-61-8
100.0
C11H11N3O
3-6-methylpyrazin-2-yl oxy aniline, benzenamine, 3-6-methyl-2-pyrazinyl oxy, 3-6-methylpyrazin-2-yloxy phenylamine, benzenamine,3-6-methyl-2-pyrazinyl oxy
CC1=CN=CC(=N1)OC2=CC=CC(=C2)N
201.229
201.23
3-[(6-Methylpyrazin-2-yl)oxy]aniline
96.5
97%
MFCD09702406
ZFDWDKWQAXTCSU-UHFFFAOYSA-N
3-(6-methylpyrazin-2-yl)oxyaniline
24229682
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
Documents

Documents

Product Certifications