missing translation for 'onlineSavingsMsg'
Learn More

3,6-Diphenyl-1,2,4,5-tetrazine, 98%

Product Code. 15720889 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15720889

Brand: Thermo Scientific Acros 453230010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 6830-78-0 | C14H10N4 | 234.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6830-78-0
Molecular Formula C14H10N4
Molecular Weight (g/mol) 234.26
MDL Number MFCD00023646
InChI Key XAUWSIIGUUMHQQ-UHFFFAOYSA-N
Synonym diphenyl-s-tetrazine, 3,6-diphenyl-s-tetrazine, 3,6-diphenyltetrazine, 1,2,4,5-tetrazine, 3,6-diphenyl, s-tetrazine, 2,6-diphenyl, s-tetrazine, 3,6-diphenyl, 3,6-diphenyl-sym-tetrazine, diphenyl-1,2,4,5-tetrazine, acmc-1baef, enamine_004961
PubChem CID 23263
SMILES C1=CC=C(C=C1)C1=NN=C(N=N1)C1=CC=CC=C1

Specifications

Melting Point 196.0°C to 198.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
IUPAC Name diphenyl-1,2,4,5-tetrazine
Formula Weight 234.26
Percent Purity 98%
Chemical Name or Material 3,6-Diphenyl-1,2,4,5-tetrazine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.